missing translation for 'onlineSavingsMsg'
Learn More
Learn More
n-Heptane, BAKER ANALYZED™, J.T. Baker™
262.20 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.
Use promo code "23268" to get your promotional price.
Click to view available options
2.5 L
Specifications
Melting Point | -90.6°C |
CAS | 142-82-5 |
Color | Max. 10 (APHA) |
Assay Percent Range | min. 99.0% |
Molecular Formula | C7H16 |
MDL Number | MFCD00009544 |
InChI Key | IMNFDUFMRHMDMM-UHFFFAOYSA-N |
SMILES | CCCCCCC |
IUPAC Name | heptane |
Molecular Weight (g/mol) | 100.21 |
Show More |
Safety and Handling
Signal Word
- Danger
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Aspiration toxicity Category 1
- Flammable liquid Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H225-Highly flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- H315-Causes skin irritation.
- H336-May cause drowsiness or dizziness.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'