missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Mouse Clusterin Quantikine ELISA Kit
Mouse Clusterin Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
Brand: R&D Systems™ MCLU00
Additional Details : Weight : 0.58200kg
Description
The Mouse Clusterin Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies mouse Clusterin/APOJ in cell culture supernates (50 ul), cell lysates (10 ul), serum (10 ul), edta plasma (10 ul), heparin plasma (10 ul), urine (10 ul).Specifications
0.78 - 50ng/mL | |
HRP | |
Natural and Recombinant Mouse Clusterin | |
12759 | |
Store the unopened product at 2° to 8°C. Do not use past expiration date. |
0.094ng/mL | |
Cell Culture Supernatant, Cell Lysates, Plasma (EDTA, Heparin), Serum, Urine | |
ELISA | |
Cell Culture Supernates (50uL),Cell Lysates (10uL),Serum (10uL),EDTA Plasma (10uL),Heparin Plasma (10uL),Urine (10uL) | |
40, 40, sulfated glycoprotein 2, Aging-associated gene 4 protein, aging-associated protein 4, apo-J, APOJ, Apolipoprotein J, CLI, CLIclusterin (complement lysis inhibitor, SP-40, CLU, clusterin, Complement cytolysis inhibitor, complement lysis inhibitor, Complement-associated protein SP-40, Ku70-binding protein 1, KUB1SGP2, MGC24903, NA1/NA2, SGP-2, SP-40, sulfated glycoprotein 2, Testosterone-repressed prostate message 2, testosterone-repressed prostate message 2, apolipoprotein J), TRPM-2, TRPM-2 TRPM2 |