missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ EnzChek™ Elastase Assay Kit

Product Code. 10257102
Click to view available options
Quantity:
600 Assays
Unit Size:
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10257102

Brand: Invitrogen™ E12056

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Fast fluorometric method for measuring elastases

The EnzChek Elastase Assay Kit provides a sensitive, convenient and fast fluorometric method for measuring elastase or other protease activity in purified enzyme systems, cell/tissue lysates or for screening inhibitors in a high-throughput format. The substrate in the EnzChek kit is BODIPY-FL-labeled DQ elastin conjugate that is highly labeled so that the fluorescence signal is quenched until enzymatic digestion yields highly fluorescent fragments.

Cell Adhesion, Cell Analysis, Cell Viability, Proliferation and Function, Cellular Imaging, Chemotaxis and Cell Migration, Enzyme and Protein Activity Assays, High-Content Screening (HCS), Protease/Peptidase Activity, Proteins, Expression, Isolation and Analysis

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in freezer -5°C to -30°C and protect from light.
Detection Method Fluorescence Intensity
For Use With (Application) Elastase Assay
Product Type EnzChek Assay Kit
Product Line EnzChek
Quantity 600 Assays
Shipping Condition Room Temperature
Substrate Properties Protein-Based Substrate
Substrate Type Elastase Substrate
Target Enzyme Elastases

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.