Learn More
Leica Microsystems Microscope Objective
Brand: Leica Microsystems 11506226
Code : 03
Additional Details : Weight : 0.80000kg
Description
The Leica DM1000 is the ideal instrument for biological, medical and clinical laboratories. The instrument can be configured to meet specific requirements by choosing from a selection of observation tubes, ergonomic modules, objectives and adjustable controls. Please note a complete system comprises the main microscope unit, an observation tube and either the brightfield or phase contrast outfit. Fluorescence outfits also require the brightfield or phase contrast outfit to be ordered.Specifications
4X | |
Objective Lens | |
HI PLAN |