missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pall Microcheck™ Beverage Monitor with GN-4 Membrane
Easy to use, versatile, bulk packaged disposable filter funnels are ideal for microbial analysis.
Brand: Pall 4762
Additional Details : Weight : 1.62450kg
Description
- Can convert to a Petri dish for culturing or easily remove the membrane for placing on separate agar dish
- Not approved for use in medical clinical surgical or other patient applications
Specifications
100 mL | |
50/Pk. | |
Gridded Filtering Funnel |
0.8 μm | |
Sterile | |
Metricel Mixed Cellulose Ester |