missing translation for 'onlineSavingsMsg'
Learn More

Methylmercury(II) chloride, standard solution in H{2}O, Methylmercury(II) chloride 1000ppm by AA

Product Code. p-4407641
Click to view available options
Quantity:
5 mL
Unit Size:
5mL
This item is not returnable. View return policy

Product Code. 11377758

Brand: Thermo Scientific Alfa Aesar 033553.AA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 115-09-3 | CH3ClHg | 251.077 g/mol

Methylmercury(II) chloride is used as a precursor for the preparation of methyl mercury acetate. It is used as a specific reagent (electron stain) for sulfhydryl groups in biological materials like protein as well as demonstration in electron microscopy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 115-09-3
Molecular Formula CH3ClHg
Molecular Weight (g/mol) 251.077
MDL Number MFCD00013592
InChI Key BABMCXWQNSQAOC-UHFFFAOYSA-M
Synonym methylmercury chloride, methylmercuric chloride, caspan, mercury, chloromethyl, methyl mercuric chloride, mercury methyl chloride, chloro methyl mercury, methyl meruric chloride, monomethyl mercury chloride, methylmercury monochloride
PubChem CID 409301
ChEBI CHEBI:28216
IUPAC Name chloro(methyl)mercury
SMILES C[Hg]Cl

Specifications

Physical Form Liquid
Quantity 5 mL
Beilstein 3600218
Solubility Information Miscible with lipid.
Formula Weight 251.08
Appearance Clear
Chemical Name or Material Methylmercury(II) chloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.