missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HMC Europe Vertical Autoclaves: HV Series
Programmable top-loading autoclaves with varying chamber volumes
Brand: HMC Europe 20002
Description
- Use steam sterilization
- Six sizes from 25 to 110 liters
- Low height, easy-to-load
- Portable — require no plumbing to operate but can be plumbed
- Complete line of baskets, inserts and cans available separately
- HV Series (dependable, hard-working, economical)
- Many upgrades available depending on your needs
Certifications
CE marked
Services
Two years on parts; One year on labor
Specifications
CE | |
100 cm | |
42 cm | |
85 L | |
25.59 in. | |
HV L85 | |
105°C to 128°C | |
71 kg | |
24 in. | |
25.98 in. |
39.37 in. | |
16.53 in. | |
22.45 gal. | |
Steam sterilization | |
65 cm | |
221°F to 262.4°F | |
156.52 lb. | |
61 cm | |
3 w | |
66 cm |