missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Javelin™ Direct-Connection Column Filters

Product Code. 11568680 Shop All Thermo Scientific Products
Change view
Click to view available options
Diameter (Metric) Outer:
2.1 mm
3 mm
4.6/4.0 mm
Unit Size:
Pack of 4
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Outer unitSize
11568680 3 mm Pack of 4
11558680 4.6/4.0 mm Pack of 4
11548680 2.1 mm Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11568680 Supplier Thermo Scientific™ Supplier No. 88700
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Protect HPLC systems with one-piece Thermo Scientific™ Javelin Direct-Connection Column Filters.

  • Direct-connection design for maximum efficiency
  • Replace entire disposable filter unit for easy changes
  • Recommended for use as dedicated filters for a column rather than the HPLC system
  • 0.0625in. CPI tip attaches directly to HPLC column inlet without tubing or wrenches
  • 0.5μm porosity
TRUSTED_SUSTAINABILITY

Specifications

Diameter (Metric) Inner 3 mm
For Use With (Equipment) HPLC Systems
Diameter (Metric) Outer 3 mm
Type Javelin Column Filter

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.