missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iodomethane, 99%, stabilized, Thermo Scientific Chemicals
29.65 GBP valid until 2024-06-30
Use promo code "21742" to get your promotional price.
29.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
Use promo code "21742" to get your promotional price.
29.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | -66.0°C |
Density | 2.2800g/mL |
Boiling Point | 42.5°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 98.5% min. (GC) |
Linear Formula | CH3I |
Refractive Index | 1.5290 to 1.5310 |
Quantity | 100 g |
Specific Gravity | 2.28 |
Solubility Information | Solubility in water: 14g/L (20°C). Other solubilities: miscible with alcohol and ether, soluble in benzene, acetone, methylenechloride, n,n-dimethylformamide |
Show More |
Safety and Handling
Product Identifier
- Iodomethane
- Danger
- Acute toxicity Category 3
- Acute toxicity Category 4
- Carcinogenicity Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H351-Suspected of causing cancer.
- H331-Toxic if inhaled.
- H335-May cause respiratory irritation.
- H312-Harmful in contact with skin.
- H301-Toxic if swallowed.
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P311-Call a POISON CENTER/doctor/ .
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'