missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Instrument Check Standard 4 For ICP/MS, SPEX CertiPrep™
Click to view available options
Quantity:
125 mL
Packaging:
Triple-leached LDPE bottle
Description
Includes
NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Analyte | Concentration |
---|---|
Mo | 10μg/mL |
Th | 10μg/mL |
U | 10μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
CAS | 7697-37-2 |
Molecular Formula | HNO3 |
MDL Number | MFCD00011349 |
InChI Key | GRYLNZFGIOXLOG-UHFFFAOYSA-N |
SMILES | O[N+]([O-])=O |
IUPAC Name | nitric acid |
Molecular Weight (g/mol) | 63.01 |
PubChem CID | 944 |
ChEBI | CHEBI:48107 |
Grade | Trace ICP/ICP-MS |
Show More |
Safety and Handling
Signal Word
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P321-Specific treatment (see on this label).
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P362+P364-Take off contaminated clothing and wash it before reuse.
- MIXTURE LIST-Contains : nitric acid
Product intended for laboratory use only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'