Learn More
ICP, ICP-MS Multi-Element Standard (23 Elements) in 5% Nitric Acid (HNO3) and 0.2% Hydrofluoric Acid (HF), Reagecon
Manufactured from ultra-pure metals or salts assayed by titration and ICP-MS prior to manufacture
Brand: Reagecon ICP23A20
Code : NEW
Additional Details : Weight : 0.01000kg Transport : UN number : 2922 Chem class : 8 Pack group : II
Description
- ISO/IEC 17025 for the testing of ICP/ICP-MS Standards using ICP-MS (INAB Ref: 264T)
- ISO/IEC 17025 for calibration of laboratory balances (INAB Ref: 265C)
- ISO/IEC 17025 for temperature calibration in the range of -196 - +1200°C (INAB Ref: 265C)
- ISO/IEC 17025 for density measurement (INAB Ref: 264T) using Oscillating U-Tube and Bingham Pycnometry
- ISO/IEC 17025 for certain complexometric titrations (INAB Ref: 264T)
- GHS/OSHA compliant, REACH etc.
- This product is produced in a highly controlled cleanroom (ISO 7) environment, using ultra-pure water (specially treated for the production of mass spectroscopy standards), triple distilled acids and triple leached pre-cleaned bottles.
- The titrimetry and spectroscopy are controlled and calibrated using separate Certified Reference Materials (CRM’s), so the product comes with two layers of traceability including NIST Standards, where available.
- All of the uncertainties of measurement for this product are calculated according to Eurachem/CITAC guidelines and reported as expanded uncertainties at the 95% confidence level.
Description Concentration Arsenic (As) 100 μg/mL(ppm) Beryllium (Be) 100 μg/mL(ppm) Calcium (Ca) 100 μg/mL(ppm) Cadmium (Cd) 100 μg/mL(ppm) Cobalt (Co) 100 μg/mL(ppm) Chromium (Cr) 100 μg/mL(ppm) Copper (Cu) 100 μg/mL(ppm) Iron (Fe) 100 μg/mL(ppm) Lithium (Li) 100 μg/mL(ppm) Magnesium (Mg) 100 μg/mL(ppm) Manganese (Mn) 100 μg/mL(ppm) Molybdenum (Mo) 100 μg/mL(ppm) Nickel (Ni) 100 μg/mL(ppm) Phosphorus (P) 100 μg/mL(ppm) Lead (Pb) 100 μg/mL(ppm) Antimony (Sb) 100 μg/mL(ppm) Selenium (Se) 100 μg/mL(ppm) Tin (Sn) 100 μg/mL(ppm) Strontium (Sr) 100 μg/mL(ppm) Titanium (Ti) 100 μg/mL(ppm) Thallium (Tl) 100 μg/mL(ppm) Vanadium (V) 100 μg/mL(ppm) Zinc (Zn) 100 μg/mL(ppm)
Specifications
Matrix: 5% HNO₃, 0.2% HF | |
HDPE Triple-leached | |
Multi-element | |
Liquid |
100 µg/mL of each element | |
100 mL | |
ICP, ICP-MS, AA, IC, XRF and other cation measurement techniques |
Safety and Handling
- ICP MULTIELEMENT STANDARD
Signal Word
- Danger
Hazard Category
- Carcinogenicity Category 1B
- Serious eye damage/eye irritation Category 1
- Oxidising liquid Category 2
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H272-May intensify fire; oxidiser.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H350i-May cause cancer by inhalation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P321-Specific treatment (see on this label).
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : nitric acid, hydrochloric aicd, hydrofluoric acid, cobalt chloride