missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molecular Probes™ Staphylococcus aureus BioParticles™ Opsonizing Reagent
Staphylococcus aureus BioParticles™ opsonizing reagent
Brand: Molecular Probes™ S2860
Code : 50
Additional Details : Weight : 0.00450kg
Description
The Staphylococcus aureus (Wood strain without protein A) BioParticles™ opsonizing agent is derived from highly purified rabbit polyclonal IgG antibodies and specifically designed to enhance the uptake of the S. aureus BioParticles™ conjugates.Specifications
Unconjugated | |
Staphylococcus aureus BioParticles™ Opsonizing Reagent | |
Staphylococcus aureus BioParticles™ Opsonizing Reagent, White, Solid, Unconjugated Dye, S. aureus Species, Store at –5 to –30°C, Used with Fluorescence Microscope, Used For Cell Structure, Cell Viability, Proliferation & Function | |
White | |
Fluorescence Microscope |
Cell Analysis, Cell Cycle, Cell Structure, Cell Viability, Endocytosis, Exocytosis, Lysosomes, Phagocytosis, Proliferation and Function, Vacuoles | |
1 Unit | |
Solid | |
-5°C to -30°C |
For Research Use Only. Not for use in diagnostic procedures.