missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ HyPerforma™ Flat-bottom LLDPE Drum, 50L, Flat-bottom outer support container for BPCs
Use this LLDPE drum with 3-D bioprocess container systems and tank liners.
Brand: Thermo Scientific™ SV50076.02
Code : 84
Additional Details : Weight : 1.00000kg
Description
Use Thermo Scientific™ HyPerforma™ Flat-bottom LLDPE Drum with 3-D bioprocess container systems and tank liners.
- Includes lid and clamps
- Available in cylindrical, conical and flat-bottom
- 4in. bottom port allows greater variety of customization to bottom port of BPC; can be used in place of drum with 1.75in. port
Specifications
Flat-bottom Outer Support Container for Bioprocess Bags, Port detail 4 and 1.75 in. | |
Top Dispense | |
45 x 53 cm (17.63 x 20.75 in.) |
50 L | |
HyPerforma™ |