missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid Solution 1M (1N), NIST Standard Solution, ready to use, for volumetric analysis, Fisher Chemical™

Catalog Number p-7073136
Click to view available options
Quantity:
2.5 L
5 L
10 L
Packaging:
Bag in box
HDPE plastic bottle
Plastic container
7647-01-0
ClH
36.46
MFCD00011324 MFCD00792839
VEXZGXHMUGYJMC-UHFFFAOYSA-N
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
313
CHEBI:17883
Cl
7647-01-0
ClH
36.46
MFCD00011324 MFCD00792839
VEXZGXHMUGYJMC-UHFFFAOYSA-N
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
313
CHEBI:17883
Cl

Hydrochloric Acid Solution 1M (1N), NIST Standard Solution, ready to use, for volumetric analysis

TRUSTED_SUSTAINABILITY
Color Colorless
Boiling Point 100°C
Packaging HDPE plastic bottle
Quantity 2.5 L
Physical Form Liquid
IUPAC Name chlorane
Formula Weight 36.46g/mol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'