missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid Solution 0.5M (0.5N), NIST Standard Solution Ready To Use, for Volumetric Analysis, Fisher Chemical™

Catalog Number p-7073138
Click to view available options
Quantity:
1 L
2.5 L
25 L
Packaging:
HDPE plastic bottle
Plastic container
7647-01-0
ClH
36.46
MFCD00011324 MFCD00792839
VEXZGXHMUGYJMC-UHFFFAOYSA-N
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
313
CHEBI:17883
hydrogen chloride
Cl
7647-01-0
ClH
36.46
MFCD00011324 MFCD00792839
VEXZGXHMUGYJMC-UHFFFAOYSA-N
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
313
CHEBI:17883
hydrogen chloride
Cl

Hydrochloric Acid Solution 0.5M (0.5N), NIST Standard Solution Ready To Use, for Volumetric Analysis

TRUSTED_SUSTAINABILITY
Color Colorless
Boiling Point 100°C
Packaging HDPE plastic bottle
Quantity 1 L
Physical Form Liquid
Formula Weight 36.46g/mol
compliance-icons
Product Identifier
  • Hydrochloric acid, 0.5M, (0.5N)
Signal Word
  • Warning
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H290-May be corrosive to metals.
Precautionary Statement
  • P234-Keep only in original container.
  • P390-Absorb spillage to prevent material damage.
Supplemental information
  • MIXTURE LIST-Contains : Hydrochloric acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'