missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrobromic Acid, ≥99.99%; Honeywell Fluka™
48 wt. % in H2O,≥99.99%
£17.86 - £55.90
Chemical Identifiers
CAS | 10035-10-6 |
---|---|
Molecular Formula | BrH |
Molecular Weight (g/mol) | 80.91 |
MDL Number | MFCD00011323 |
InChI Key | CPELXLSAUQHCOX-UHFFFAOYSA-N |
Synonym | hydrogen bromide, hydrobromic acid, bromwasserstoff, broomwaterstof, bromowodor, acido bromidrico, acide bromhydrique, anhydrous hydrobromic acid, bromowodor polish, hydrobromide |
PubChem CID | 260 |
ChEBI | CHEBI:47266 |
SMILES | Br |
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
10035-10-6 | |
80.91 | |
CPELXLSAUQHCOX-UHFFFAOYSA-N | |
260 | |
Br |
BrH | |
MFCD00011323 | |
hydrogen bromide, hydrobromic acid, bromwasserstoff, broomwaterstof, bromowodor, acido bromidrico, acide bromhydrique, anhydrous hydrobromic acid, bromowodor polish, hydrobromide | |
CHEBI:47266 |
Specifications
Acidic | |
10035-10-6 | |
-11°C | |
1.49g/mL at 25°C | |
320psi (21.1°C), 8mmHg (25°C) | |
MFCD00011323 | |
3587158 | |
Completely miscible in water | |
Br | |
80.91 | |
CHEBI:47266 | |
≥99.99% |
Yellow | |
48 wt.% in water | |
126°C | |
Stinging | |
BrH | |
UN1788 | |
hydrogen bromide, hydrobromic acid, bromwasserstoff, broomwaterstof, bromowodor, acido bromidrico, acide bromhydrique, anhydrous hydrobromic acid, bromowodor polish, hydrobromide | |
CPELXLSAUQHCOX-UHFFFAOYSA-N | |
hydrogen bromide | |
260 | |
80.91g/mol | |
Hydrobromic acid |
Safety and Handling
P260-P280-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338
missing translation for 'dotInformation' : Class 8/ PGII