missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ Characterized Fetal Bovine Serum, Australian Origin
Conformation Tested According to the European Agency for the Evaluation of Medicinal Products (EMEA)
£1147.50 - £2177.70
Specifications
Country of Origin | Australia |
---|---|
Form | Serum |
Product Type | Fetal Bovine Serum |
Sterilization Method | Sequential Triple-filtered, 100 nm |
Product Line | HyClone™ |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16635775
|
Cytiva
SH30545.03HI |
500 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16645775
|
Cytiva
SH30545.04HI |
1000 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16655775
|
Cytiva
SH30545.04NI |
1000 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16625775
|
Cytiva
SH30545.03 |
500 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Sourced from Australian abattoirs approved by USDA for export and inspected by the Australian Department of Agriculture (DA)
- Australian methods of animal husbandry exhibit excellent animal nutrition and healthcare
- Animal disease control and management in Australia is easier compared to most areas of the world due to isolation
- Filtered through triple 100 nm pore size-rated filters
- Standard endotoxin and hemoglobin specifications
- Subjected to moderate biochemical analyses
Specifications
Australia | |
Fetal Bovine Serum | |
HyClone™ |
Serum | |
Sequential Triple-filtered, 100 nm | |
European Agency for the Evaluation of Medicinal Products (EMEA) Conforming Tests |