Learn More
Rotronic™ Humidity standards
For on-site calibration and adjustment of ROTRONIC probes with handheld instrument
£124.78 - £132.98
Specifications
Certificate of Analysis | SCS traceable calibration certificate, textile pad, calibration instruction |
---|---|
For Use With (Equipment) | Rotronic Instrument |
Product Code | Brand | Packaging | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Packaging | Price | Quantity & Availability | |||||
10781977
|
Rotronic™
EA05-SCS |
0.05 |
£124.78
5 ampoules |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
13114967
|
Rotronic™
EA35-SCS |
0.35 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
With the humidity standards, a calibration device and HW4 software, running on a PC this is easily achieved.
- Ampoules contain unsaturated salt solutions
- Simple and safe on-site calibration method
- Unlimited storage life
Specifications
SCS traceable calibration certificate, textile pad, calibration instruction | |
Rotronic Instrument |