Learn More
Invitrogen™ Human SCNN1B (aa 619-640) Synthetic Peptide
Synthetic Peptide
Brand: Invitrogen™ PEP089
Code : LE
Additional Details : CAS Number : 69-74-9 Weight : 0.01000kg
Description
Human beta-ENaC (residues 619-640) Synthetic Peptide, PEP-089, from Invitrogen.
PEP-089 is a 23 amino acid synthetic peptide whose sequence corresponds to amino acids 619-640 of the human beta-ENaC with an N-terminal cysteine added for carrier protein conjugation. The sequence of this peptide is (amino to carboxy terminus): C N (619)-Y-D-S-L-R-L-Q-P-L-D-V-I-E-S-D-S-E-G-D-A-I (640)This peptide may be used for neutralization and control experiments with the polyclonal antibody that reacts with this product and human beta-ENaC, catalog # PA1-921. Using a solution of peptide of equal volume and concentration to the corresponding antibody will yield a large molar excess of peptide (∼70-fold) for competitive inhibition of antibody-protein binding reactions.Reconstitute with 0.1 ml of distilled water.Specifications
6338 | |
purified | |
Human SCNN1B (aa 619-640) | |
-20°C. Avoid freeze-thaw cycles. | |
Lyophilized | |
Control | |
Yes | |
Peptide | |
RUO - research use only |
Human | |
P51168 | |
0.5 mg/mL | |
Synthetic Peptide | |
peptide with no preservative | |
SCNN1B | |
No preservative | |
50 μg | |
C N(619)-Y-D-S-L-R-L-Q-P-L-D-V-I-E-S-D-S-E-G-D-A-I(640) |