missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human LIF Synthetic Peptide

Synthetic Peptide

Brand:  Invitrogen™ PEP1236

Code : NEW

Additional Details : Weight : 0.01000kg

Product Code. 15235239

  • £112.00 / 50µg
Estimated Shipment: 11-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

LIF Synthetic Peptide, PEP-1236, from Invitrogen.

This peptide corresponds to 16 amino acids near the center of human LIF.PEP-1236 can be used as a blocking peptide with polyclonal antibody PA5-21122.
Specifications

Specifications

3976
purified
Human LIF
-20°C
Liquid
Blocking Assay, Control
No
Peptide
RUO - research use only
Human
P15018
200 μg/mL
Synthetic Peptide
PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
LIF
0.02% sodium azide
50 μg
PBS
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers