Learn More
Invitrogen™ Human CaMKI (aa 1-18) Synthetic Peptide
Synthetic Peptide
Brand: Invitrogen™ PEP134
Code : LE
Additional Details : CAS Number : 69-74-9 Weight : 0.01000kg
Description
Human CaMK1 (residues 1-18) Synthetic Peptide, PEP-134, from Invitrogen.
PEP-134 is an 18 amino acid synthetic peptide whose sequence corresponds to amino acid residues 1-18 from human CaMKI protein. The sequence of this peptide is (amino to carboxy terminus): A (1)-V-E-G-P-R-W-K-Q-A-E-D-I-R-D-I-Y-D (18).This peptide may be used for neutralization and control experiments with the polyclonal antibody that reacts with this product and human CaMKI protein, catalog # PA1-543. Using a solution of peptide of equal volume and concentration to the corresponding antibody will yield a large molar excess of peptide (∼70-fold) for competitive inhibition of antibody-protein binding reactions.Reconstitute with 0.1 ml of distilled water.Specifications
8536 | |
purified | |
Human CaMKI (aa 1-18) | |
-20°C. Avoid freeze-thaw cycles. | |
Lyophilized | |
Control | |
Yes | |
Peptide | |
RUO - research use only |
Human | |
Q14012 | |
0.5 mg/mL | |
Synthetic Peptide | |
peptide with no preservative | |
CAMK1 | |
No preservative | |
50 μg | |
A(1)-V-E-G-P-R-W-K-Q-A-E-D-I-R-D-I-Y-D(18) |