missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HiPrep™ Phenyl HP 16/10 Column

Designed for preparative hydrophobic interaction chromatography (HIC) with good resolution

Brand:  Cytiva 29018184

Code : GE

Additional Details : Weight : 0.11100kg


Product Code. 14891601

  • £671.00 / Each
Estimated Shipment: 11-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Aromatic HIC medium for intermediate purification and polishing
  • Provides fast, easy high-capacity separations in a convenient format
  • Medium is based on a matrix of highly cross-linked agarose beads, which have been derivatized with phenyl groups

Specifications

Specifications

0.6 in.
4°C to 30°C, 0.2M Sodium Acetate in 20% Ethanol
20 mL
1 to 5 mL/min.
16 mm
1 x 20 mL
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

SDS
Documents

Documents

Certificates
Special Offers

Special Offers