Learn More
FTM Technologies™ HI5VP Extraction Booths
Used for filtration needs where a channelled space is needed. FTM Technologies™ HI5VP Extraction Booths have shaped, shatterproof glass measuring 6mm thick.
Brand:  FTM Technologies™ HI5VP
Additional Details : Weight : 31.00000kg
Product Code. 11758054
  • £3315.00 / Each
Estimated Shipment: 06-06-2024
to see stock.
Description
Description
  • Front of booth entirely open
  • Applications include chemical handling, spraying, pulverization and varnishing
  • Also suitable for cleaning solvents, tubs, or machines producing a large amount of fumes
  • Can be used as recycling or ductless - filtration with active carbon or dust filter
  • Can also be used with external evacuation (filter for evacuation plus exhaust duct)
Specifications
Specifications
800 x 700 x 700 mm
50 Hz
33 kg
50 to 450 m3/h
230 V
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products