Learn More
Excel Healthcare Ltd™ Helix Test Strips
These tests are easy to complete and record, giving full performace traceability of your vacuum steriliser
Brand: Excel Healthcare Ltd™ 188
Additional Details : Weight : 1.00000kg
Description
- Excel Healthcare Has A Range Of Testing Products That Are Compatible With Our Range Of Steam Sterilisers
- Manual Testing Of Steam Sterilisers Is Now Considered An Essential Requirement And Must Be Performed By The Operator On A Daily Basis
- Htm 01-05 States That: Daily Tests Must Be Performed By The Operator Or User And If The Steriliser Fails To Meet Any Of The Test Requirements The Steriliser Must Be Withdrawn From Service And Advice Sought From The Manufacturer Or Service Provider
- The Daily Tests Consist Of Steam Penetration Test (Vacuum Sterilisers Only) And Automatic Control Test. These Tests Should Be Dated, Signed And Recorded In The Steriliser Log Book
- These Tests Are Easy To Complete And Record, Giving Full Performance Traceability Of Your Vacuum Steriliser
- Efficient Testing Device
- Detects Atleast One Vacuum Is Pulled Per Cycle
- To Be Inserted Into Helix Testing System
Specifications
Conform to EN 867 – 1, ISO 11140 – 1, Class 2 | |
Helix Test Strips | |
Inks are non-toxic and the adhesives do not contain latex | |
200 |
For Use with Helix Test System | |
Self-adhesive helix strips | |
1 pack of 200 |