missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Grade 602 Medium Filtering Special Filter Paper

Suitable for all works in laboratories. Fisherbrand™ 602 Medium Filtering Special Filter Paper is used in filtration of various products, pharmaceutical wrapping (pills, powders, dressing bandages and tablets).

Brand:  Fisherbrand™

Code : A0

Additional Details : Weight : 0.24300kg

 View more versions of this product

Product Code. 11754566

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Description

Description

  • Color: Vellum grey
Specifications

Specifications

Folded Sheet
602
520 x 420mm
420 mm
All works in laboratories - filtration of various products, pharmaceutical wrapping (pills, powders , dressing bandages and tablets)
25PK
520 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers