missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gilson™ PIPETMAN M Single Channel Pipets
Shop All Gilson Products
Click to view available options
Volume (Metric):
0.5 to 10 μL
2 to 20 μL
20 to 200 μL
5 to 100 μL
Unit Size:
Each
Description
An ergonomic pipetting solution with GLP features that each lab needs
- Motorized piston with adjustable speed that is compatible with varying solution densities
- Lockable volume to prevent inaccuracies
- Color-coded push-button corresponds to matching tip packs
- Service alert reminders
- Unique tip holder design allows for a secure fit of most brands of tips
- Features four essential pipetting modes – reverse, mix, repetitive and pipetting – to best fit your application
- Pipette can be used while charging eliminating bottlenecks and downtime
- Power carrousel conveniently holds and charges up to 5 PIPETMAN M pipettes at once, charging 80% of its full capacity in less than one hour
Specifications
Specifications
| For Use With | D-10, DL10, DF10ST, DFL10ST Pipette Tip |
| Number of Channels | 1 |
| Product Line | Pipetman M |
| Type | Pipette |
| Volume (Metric) | 0.5 to 10 μL |
| For Use With (Equipment) | D10, DL10, DF10ST, DFL10ST pipette tip |
| Includes | Fully motorized piston, color coded push button, universal tip holder |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction