missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molecular Probes™ Fura-2, Pentapotassium Salt, cell impermeant
Fura-2, Pentapotassium Salt, cell impermeant
Brand: Molecular Probes™ F1200
Code : 50
Additional Details : Weight : 0.00454kg
Description
Fura-2, pentapotassium salt is an intracellular calcium indicator that is ratiometric and UV light-excitable. This water-soluble salt form is useful for intracellular loading by microinjection, infusion from patch pipette or uptake induced by our Influx pinocytic cell-loading reagent (I14402).Specifications
Yellow | |
–5 to –30°C | |
Fluorescent | |
Fluorescence Microscope | |
Solid |
60 to 100% | |
Fura-2 Pentapotassium Salt, 1mg, Yellow, Solid, 831.99 Molecular Weight, C29H22K5N3O14 Molecular Formula, Fluorescent Detection, Cell-imPermanent, –5 to –30°C Storage, ≥95% at 254nm HPLC Purity, Use with Fluorescence Microscope | |
Cell Viability, Proliferation and Function, Cellular Imaging, Target and Lead Identification and Validation | |
335nm | |
1mg |
For Research Use Only. Not for use in diagnostic procedures.