missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ohaus™ Frontier™ 5000 Series Micro-Centrifuge
Dependable benchtop micro-centrifuge, ideal for high-speed laboratory applications.
£2040.00 - £4460.00
Specifications
Max. RCF | 21,953 x g |
---|---|
Display | Backlit LCD |
Safety Features | CEFCC |
Maximum Speed Range | 200 to 15,200 rpm |
Rotor Included | No |
Product Code | Brand | Dimensions (L x W x H) Exterior | Net Weight | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions (L x W x H) Exterior | Net Weight | Price | Quantity & Availability | ||||
15876752
|
Ohaus™
30130866 |
393.00 x 287.00 x 292.000 mm | 17.000 kg |
£2040.00
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15886752
|
Ohaus™
30130868 |
561.00 x 287.00 x 292.000 mm | 35.000 kg |
£4460.00
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
- High-speed operation for a range of essential lab applications, such as DNA/RNA preparation, ultrafiltration, and more.
- Biocontainment rotors enable research involving hazardous samples.
- Engineered with convenience of operation in mind, these centrifuges feature an intuitive interface, with all key parameters immediately accessible for observation and adjustment.
- The compact design saves valuable bench space in the lab.
Specifications
21,953 x g | |
CEFCC | |
No | |
Frontier 5000 Series Micro |
Backlit LCD | |
200 to 15,200 rpm | |
10 sec to 99 h 59 min 59 secContinous | |
230 V |