Learn More
Faster™ FlowFAST V 15 Vertical Laminar Flow Cabinet
FlowFAST V are positive pressure ISO Class 3 vertical laminar flow cabinets, which are an economical alternative to cleanrooms. They are capable of providing uncontaminated, particle-free and sterile conditions for materials in the workspace.
Brand:  Faster™ F00001150000
Additional Details : Weight : 250.00000kg
Product Code. 16228470
  • £8185.00 / Each
Estimated Shipment: 02-09-2024
to see stock.
Description
Description
Ergonomic Design: The angled sloping (7°) front hinged safety-glass provides optimum visibility of all objects placed in the interior workspace. The frontal opening is set to 200 mm. The whole front of the cabinet can also be opened upwards as it is hinged on the top – in order to allow easy access for complete and effective cleaning and decontamination.
Real Laminar AirFlow: Frontal screen 7° sloped as well as back side wall to convey in unidirectional pattern the air flow. As a consequence, the front and back panels are parallel one with the other and there is the real presence of Laminar AirFlow in the whole working area.
ECS™ Eco Controling System: The new ECS™ microprocessor employs the latest innovative methods of integrated management of all principal functions of ventilation and filtration - self-regulating all the main filtration and ventilation system components - compensating for declining pressure drops and restoring power balance. Combining the use of AC motor-blower and certified low pressure-drop filters, the new ECS™ controlling system optimize power consumption, reducing CO2 emissions into the environment.
Anti Bacterial Coating: Exclusive Dupont™ ALESTA™ anti-bacterial 'Ag+ cations-based solution', capable to prevent microbial contamination of surfaces thereby inhibiting long term surface growth. Work surface in stainless steel AISI 304L. Cold rolled steel structure.
High Level Lighting: The safety glass side-windows with the ideal positioning and sizing of the light-system provide the highest level of luminosity to the work area.
Easy handling and maintenance: The cabinet can pass through standard 800 mm wide door openings. In fact, the overall depth of the cabinet is 760 mm. All service operations are available from the front of the cabinet. Useful snap hinges keep in open position the frontal tempered glass increasing the height of the frontal opening. This solution allows the entrance of devices (such as microscopes) beneath laminar flow.
Electrical sockets fitted as standard in each size model: one (for size 12) or two (for size 15 and 18).
Specifications
Specifications
H14 Hepa
Epoxy-coated Metal