Learn More
Faster™ FlowFAST H 15 Safety Cabinet with Stainless Steel Work Surface
FlowFAST H are positive pressure ISO Class 3 horizontal laminar flow cabinets capable of providing uncontaminated, particle-free, and sterile conditions for materials in the workspace. More economical to cleanrooms.
Brand:  Faster™ F00000210000
Additional Details : Weight : 1.00000kg
Product Code. 16298460
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
Ergonomic Design and best space utilization: The angled sloping front shape provides optimum visibility of all objects placed in the interior workspace. The whole front of the cabinet is totally open in order to guarantee the best space available inside the working area and this solution allows the entrance of big devices (such as microscopes) beneath laminar flow.
Full laminar flow in the whole volume area thanks to the total covering of the rear surface by H 14 HEPA/ULPA filter (without any frame/items obstacles).
H 14 HEPA/ULPA filter with air diffusor which guarantees the ideal spread of laminar air flow in each zone of the working area.
ECS™ Eco Controling System: The new ECS™ microprocessor employs the latest innovative methods of integrated management of all principal functions of ventilation and filtration - self-regulating all the main filtration and ventilation system components - compensating for declining pressure drops and restoring power balance. Combining the use of AC motor-blower and certified low pressure-drop filters, the new ECS™ controlling system optimize power consumption, reducing CO2 emissions into the environment.
Anti Bacterial Coating: Exclusive Dupont™ ALESTA™ anti-bacterial 'Ag+ cations-based solution', capable to prevent microbial contamination of surfaces thereby inhibiting long term surface growth. Work surface made of stainless steel AISI 304 L.
High Level Lighting: The big size safety glass side-windows along with the total opening of the frontal aperture, provide the highest level of luminosity to the work area.
Easy handling and maintenance: The cabinet has been conceived to be easily demountable and remountable in order to pass through standard 800 mm wide door openings.
Electrical socket fitted as standard in each size model.
Specifications
Specifications
H14 Hepa
Stainless Steel