Learn More
Erlab™ Fixed Benchcap
Erlab™ Fixed Benchcap is a fixed metal support cabinet for use with captairFlex filtration hoods.
Brand:  Erlab™ I0518000302
Code : NEW
Additional Details : Weight : 0.80000kg
Product Code. 15442414
  • £1245.50 / Each
Estimated Shipment: 04-09-2024
to see stock.
Description
Description
  • Benchcap
  • For use with CaptairFlex XLS 483
  • Fixed metal support cabinet
  • Equipped with 4 cylinders for leveling the unit
Specifications
Specifications
Fixed Benchcap
Fixed Bench
4 cylinders for leveling
Metal
CaptairFlex XLS 483