Learn More
Fisherbrand™ Finnpipette™ II Digital Multichannel Pipetters
Advanced pipetting performance in a lightweight, ergonomic design
Brand: Fisherbrand™
Code : 46
Additional Details : Weight : 0.01000kg
Description
- 8- and 12-channel pipetters, ideal for multiplate applications
- Minimum force allows tips to be ejected with just a gentle push
- Reverse pipetting mode allows pipetting of viscous and surface-active solutions
- Made from inert, biologically inactive polymers
- Easy to disassemble for routine maintenance and cleaning
- Serial number and calibration certificate included
- Easy, in-lab calibration
Superior Design
- Rounded hand grip and modified grippy finger rest ensure secure grip and correct pipetting angle; help minimize hand fatigue
- Large digital display is easy to read, even when pipetting
- Large color-coded knob makes it easy to set and identify desired volume; fast-dial system allows 0.1μL fine adjustment
- Separate soft-touch tip ejector relieves hand stress, prevents accidental detipping
- Super blowout feature on micro models ensures highest level of accuracy and precision at low-volume settings
Order Info
For optimum performance, use Fisherbrand SureOne™ Pipet Tips or other universal fit pipet tips.
Services
Three years
Specifications
Autoclavable | |
12 | |
Digital Pipetter | |
5μL |
Yellow | |
Finnpipette™ II | |
50 to 300 μL | |
3 Years |