Learn More
Lenz Laborglasintrumente Extraction Thimbles
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
£42.00 - £54.00
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
10274051
|
Lenz Laborglasintrumente
05431203 |
250 mL |
£54.00
Each |
Estimated Shipment: 19-06-2024
Log in to see stock. |
|||||
10243911
|
Lenz Laborglasintrumente
05431202 |
100 mL |
£45.00
Each |
Estimated Shipment: 19-06-2024
Log in to see stock. |
|||||
10644392
|
Lenz Laborglasintrumente
05431201 |
70 mL |
£42.00
Each |
Estimated Shipment: 19-06-2024
Log in to see stock. |
|||||
Description
- Extraction thimbles
- Glass insert with bores for solids
- Made of DURAN™ tubing
Specifications
Borosilicate Glass (3.3) |