Learn More
Bellingham Stanley™ Eclipse Professional Grade Optical Hand-Held AntiFreeze Refractometer
Brand:  Bellingham Stanley™ 45-44
Code : 04
Additional Details : Weight : 0.01000kg
Product Code. 12333529
  • £262.88 / Each
Estimated Shipment: 15-07-2024
to see stock.
Add to basket
Specifications
Eclipse Professional Grade Optical Hand Held Refractometer
0°C to -40°C Antifreeze Protection, 1.1 to 1.35 Battery Acid Specific Gravity
‘On the spot’ determination of Refractive Index (RI) of liquids or the concentration of solutions
IP65