Learn More
STEMCELL Technologies EasySep™ Red Blood Cell Lysis Buffer
Red blood cell lysis buffer, 10X concentrate
Brand:  STEMCELL Technologies 20110
Additional Details : Weight : 0.05000kg
 View more versions of this product
Product Code. 17108021
  • £70.20 / 10mL
Estimated Shipment: 21-05-2024
to see stock.
Description
Description
  • EasySep™ Red Blood Cell Lysis Buffer is for use as directed with EasySep™ or RoboSep™ Whole Blood kits for the lysis of red blood cells prior to separation from whole blood
Specifications
Specifications
Chimerism; HLA; Immunology
10 mL