missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences DURAN™ Aspirator Laboratory Bottle, GL 45 Thread Neck, GL 32 Thread Side Arm
Click to view available options
Capacity (Metric):
10,000 mL
1000 mL
2000 mL
5000 mL
Description
Aspirator laboratory bottle with screw closure, complete with PTFE key stopcock and GL 45 red PBT cap.
- Supplied ready to use
- DURAN™ glass and PTFE key stopcock outlet
- Made of virtually inert, chemically resistant DURAN™ glass
- Fully compatible with all the GL 45 threads and therefore highly versatile
- Tabulator with GL 32 thread
- Easy-to-read scale with graduation marks to estimate the content
- Fired-on, highly durable white ceramic
Specifications
Capacity (Metric) | 2000 mL |
For Use With (Application) | Storage and dispensing of liquids and reagents |
Material | Borosilicate Glass (3.3), Silicone, PTFE |
No. per Pack | 1/Pk. |
Neck Finish | Thread Size GL 45 |
Autoclavable | Autoclavable |
Diameter (Metric) | 136 mm |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'