Learn More
Trajan Scientific and Medical™ Dual Gauge Spare Needles
Replacement needle for Agilent Technologies Autosampler Syringes
Brand: Trajan Scientific and Medical™ 037730
Code : 13
Additional Details : Weight : 0.20000kg
Description
- Dual Gauge: 23, 26
- Gas Tight
- 0.63/0.47mm OD
Specifications
23 and 26 G | |
0.47 to 0.63 mm | |
10 μL | |
Syringes (10μL) |
0.47 to 0.63 mm | |
Replacement Needle | |
Cone |