Learn More
Nickel Electro™ Draining Syphon
Nickel Electro™ Draining Syphon allows water baths to be emptied quickly.
Brand:  Nickel Electro™ BX0616
Code : 03
Additional Details : Weight : 0.01000kg
Product Code. 11419539
  • £57.28 / Each
Estimated Shipment: 06-06-2024
to see stock.
Add to basket
Description
Description
  • Water temperature must be below 45°C before use
  • Allows the easy and quick emptying of water baths, immerse into water bath, squeeze the bellow and point the outlet to a bucket or sink
Specifications
Specifications
NE4-8 D/CT, NE1B-28, NE1-2.5 E, NE4-28 D, NE4-22 T Stirred Digital Clear Water Baths
Below 45°C
Draining Syphon