Learn More
Fisherbrand™ Deuterium Lamps
Designed for compatibility with a variety of instruments. Fisherbrand™ Deuterium Lamps are individually tested, certified, and guaranteed to the same specifications as OEM.
Brand: Fisherbrand™
Additional Details : Weight : 0.50000kg
Description
- Produced in state of the art manufacturing facilities approved by lamp manufacturers
- Full traceability of manufactured products
- Lamps available for all major instruments
Specifications
Deuterium Lamp | |
400/185 nm | |
Agilent 1290 DAD |
UV | |
0.5 mm | |
1 Ea. |