Learn More
Fisherbrand™ Deuterium Lamps
Designed for compatibility with a variety of instruments. Fisherbrand™ Deuterium Lamps are individually tested, certified, and guaranteed to the same specifications as OEM.
Brand: Fisherbrand™
Code : NEW
Additional Details : Weight : 0.25000kg
Description
- Produced in state of the art manufacturing facilities approved by lamp manufacturers
- Full traceability of manufactured products
- Lamps available for all major instruments
Specifications
Deuterium Lamp | |
Dionex PDA-100, PDA-3000, AD-25 |