missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ DAB Quanto Detection System
Provides a strong brown staining and can be used after reconstitution for up to 1 week at room temperature and up to 2 weeks at 2° to 8°C.
£291.00 - £1960.00
Specifications
Chromagen Type | DAB Quanto Chromagen |
---|---|
Substrate Type | Dab Quanto Subrate |
Alert:
DAB Quanto Chromogen contains Diaminobenzidine (DAB) in buffer. DAB is a suspected carcinogen. Do not pipet by mouth. Avoid contact with skin and eyes. Reagent is acidic and can cause burns if skin contact occurs. Handle with care and dispose of according to regulations. DAB Quanto Substrate contains Hydrogen Peroxide in buffer. Avoid contact with skin and eyes.
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
12623957
|
Epredia™
TA-060-QHDX |
60 mL DAB Quanto Substrate, 2mL DAB Quanto Chromagen |
£291.00
60mL |
Estimated Shipment: 23-05-2024
Log in to see stock. |
|||||
12693967
|
Epredia™
TA-125-QHDX |
125mL DAB Quanto Substrate, 4mL DAB Quanto Chromagen |
£580.00
125mL |
Estimated Shipment: 23-05-2024
Log in to see stock. |
|||||
15260715
|
Epredia™
TA-999-QHDX |
1L |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
DAB Quanto has been validated in combination with both UltraVision LP and UltraVision Quanto and provides four major benefits.- Strong staining—ideal to quantifying cells and protein expression
- Less mixing—reducing personnel time and stress
- Less disposal—cuts down waste
- Lower risk of mixing errors—supports higher staining consistency
- Supplied in a two-component form consisting of a DAB Quanto Chromogen and DAB Quanto Substrate
- Standard working dilution is 30μL of DAB Quanto Chromogen per 1mL of DAB Quanto Substrate
- Use of liquid components reduces some risks associated with the handling of powders or tablets
Specifications
DAB Quanto Chromagen | |
Dab Quanto Subrate |