Learn More
Arctiko™ CRYO 230 Cryogenic Freezer
The CRYO range features our highly reliable temperature technology comprised of two independent systems to offer double security.
Brand: Arctiko™ DAI 0198
Additional Details : Weight : 563.00000kg Transport : UN number : 2794 Chem class : 8
Description
The CRYO range features the highly reliable TRUE DUAL™ technology comprised of two independent systems, offering double security. In the unlikely event that one cooling system should fail, the second cooling system will maintain the freezer at -130°C. The CRYO 170 offers a wide variety of research and long-term storage applications for low temperature scientific experiments, preservation of cells, DNA, bone material, bacteria, etc. With its superior cooling precision and features, the CRYO 170 is the perfect choice for testing special material, making it ideal for hospitals, university laboratories, military industry or sanitation stations.
- Independent TRUE DUAL™ technology for double security
- Low energy consumption
- Low noise level
- Visual and acoustic alarms
- Digital display
- Temperature chart recorder
- Battery back up
- Porthole for the external temperature probe
- 100% CFC and HCFC free
- Prepared for LN2 Backup (Optional)
- Depth with detachable airflow management and noise reduction system: 1135 mm