missing translation for 'onlineSavingsMsg'
Learn More

Corning™ HTS Transwell™ -96 Well Permeable Support System

Boasts membrane area for cell growth or migration of 0.143cm2. Corning™ HTS Transwell™ 96-Well Permeable Support System is a microplate support system for an array of 96 wells with membrane inserts connected by a rigid, robotics-friendly tray that enables all 96 inserts to be handled as a single unit.

Brand:  Corning™ 3386

Code : 67

Additional Details : Weight : 0.95254kg

 View more versions of this product

Product Code. 10077792

  • £667.37 / Pack of 8
Estimated Shipment: 04-10-2024
to see stock.
Add to basket

Description

Description

  • The HTS Transwell-96 permeable support has an array of 96-wells with membrane inserts connected by a rigid, robotics-friendly tray that enables all 96 inserts to be handled as a single unit.
  • Choice of either polyester (PET) membrane (0.4 μm, 1.0 μm pore sizes) or polycarbonate (PC) membrane (0.4 μm, 3.0 μm, and 5.0 μm pore sizes)
  • 0.143 cm2 membrane area per well, providing 20% to 50% more surface area for cell growth than other commercially available systems
  • Large apical and basolateral access ports allow efficient media sampling and facilitate automated or manual access.
  • Optimized for automation, with multichannel feeder ports, improved gripping surface, and standard bar codes
  • The reservoir plate allows for simultaneous feeding of 96 wells and comes with a removable media stabilizer to reduce the risk of spills during handling
  • The receiver plate isolates each well to enable 96 individual assays.
  • Sterile
  • The HTS Transwell-96 systems (0.4 μm PC, 0.4 μm PET, and 1.0 μm PET) are packaged with the 96-well insert plate in a reservoir plate and includes the 96-well receiver plate with lid.
  • The HTS Transwell-96 plates (3.0 and 5.0 μm PC, 8.0 μm PET) are packaged with the 96-well insert plate in the 96-well receiver plate with lid. Reservoir plates may be purchased separately.
Specifications

Specifications

Standard
TC-treated polycarbonate
3 μm
Polycarbonate
0.23 mL
Flat Bottom (F-well)
8/Cs.
4.26 mm
0.143 cm2
8 x 12
Clear
96
Sterile
HTS Transwell™-96 permeable support insert, receiver plate (96 wells for growth or assay steps) and 1 sterile lid
4/Pk.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers