Learn More
Corning™ Synthemax™ II-SC Substrate
Self-coating synthetic surface for stem cells
Brand: Corning™ 3535
Code : NEW
Additional Details : Weight : 0.00450kg
Description
Easy-to-use, self-coatingmaterial for creating a unique synthetic surface that mimics the natural cell environment. Surface supportsthe adhesion and expansion of stem cells in their undifferentiated state and directed differentiation into specialized cell types. In addition, it offers a synthetic, xeno-free alternative to biological coatings and/or feeder cell layers used in hES and iPS cell culture.
Allows for the scalable multi-passage expansion of pluripotent stem cells in defined media with:
- Stable doubling time
- Phenotypicmarker expression (Oct4, SSEA4)
- Retention of pluripotency
- Retention of normal karyotype
- Easy-to-use
- Able to coat plastic and glass surfaces
- Lot-traceable
- Gamma sterilized
- Quality tested for lot-to-lot consistency
Specifications
Creating a unique synthetic surface that mimics the natural cell environment | |
1 mg/mL Synthemax II SC stock solution, sterile water, 0.025 mg/mL final volume of Corning™ Synthemax II SC working solution | |
10 mg |
Synthemax™ II-SC Substrate | |
-20°C |