missing translation for 'onlineSavingsMsg'
Learn More

Chromone-2-carboxylic acid, 97%

Product Code. p-7021278
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11430044

Brand: Thermo Scientific Alfa Aesar A13552.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 4940-39-0 | C10H6O4 | 190.154 g/mol

Chromone-2-carboxylic acid is used in the cyclic form 3?-oxopyrazolidino[4?,5??2, 3]-chroman-4-one, gives the azide of chromone-2-carboxylic acid, the curtius rearrangement of which is used to synthesize a number of 2-acylaminochromones and it is an orally active antiallergic agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chromone-2-carboxylic acid is used in the cyclic form 3′-oxopyrazolidino[4′,5′∶2, 3]-chroman-4-one, gives the azide of chromone-2-carboxylic acid, the curtius rearrangement of which is used to synthesize a number of 2-acylaminochromones and it is an orally active antiallergic agent.

Solubility
Soluble in alcohol, water, DMSO, hot methanol, and ethanol.

Notes
Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4940-39-0
Molecular Formula C10H6O4
Molecular Weight (g/mol) 190.154
MDL Number MFCD00006838
InChI Key RVMGXWBCQGAWBR-UHFFFAOYSA-N
Synonym chromocarb, chromone-2-carboxylic acid, 4-oxo-4h-chromene-2-carboxylic acid, 4-oxo-4h-1-benzopyran-2-carboxylic acid, atremon, chromonecarboxylic acid, lp-1, chromocarb inn:dcf, 2-chromonecarboxylic acid, chromocarbe inn-french
PubChem CID 2741
IUPAC Name 4-oxochromene-2-carboxylic acid
SMILES C1=CC=C2C(=C1)C(=O)C=C(O2)C(=O)O

Specifications

Melting Point ∼260°C (decomposition)
Quantity 5 g
Beilstein 146442
Merck Index 14,2237
Solubility Information Soluble in alcohol,water,DMSO,hot methanol,and ethanol.
Formula Weight 190.15
Percent Purity 97%
Chemical Name or Material Chromone-2-carboxylic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.