missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chloroform, Contains ethanol as stabilizer, meets analytical spec. of DAB9, BP, 99-99.4% (GC), Honeywell Riedel-de Haën™
Click to view available options
Quantity:
1 L
2.5 L
40 kg
4 x 2.5 L
6 x 1 L
250 kg
Description
- Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
- It is the leading brand offering High-Purity Solvents for dedicated applications.
Specifications
Density | 1.492g/mL at 25°C, 1.479 to 1.489g/mL at 20°C |
Quantity | 250 kg |
Vapor Pressure | 160mmHg (20°C) |
UN Number | 1888 |
Beilstein | 1731042 |
IUPAC Name | trichloromethane |
Formula Weight | 119.38g/mol |
Percent Purity | 99 to 99.4% (GC) |
Chemical Name or Material | Chloroform, Meets analytical specification of DAB9, BP |
Safety and Handling
Product Identifier
- Chloroform
- Danger
- Acute toxicity Category 4
- Acute toxicity Category 3
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Carcinogenicity Category 2
- Reproductive toxicity Category 2
- Specific target organ toxicity Category 3
- Specific target organ toxicity Category 1
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H331-Toxic if inhaled.
- H336-May cause drowsiness or dizziness.
- H351-Suspected of causing cancer.
- H361d-Suspected of damaging the unborn child.
- H372-Causes damage to organs.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'