missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Asecos™ CF-CLASSIC Safety Storage Cabinet
For the safe and approved indoor storage of different hazardous substances in one safety storage cabinet.
Brand: Asecos™ 30796
This item is not returnable.
View return policy
Description
- Robust construction and longevity – sturdy sheet steel construction
- No unauthorized use – doors lockable with cylinder locking, integrated in the turning handle
- Ventilation – ventilation openings in the door, ready for connection (DN 75) to a technical exhaust system
- Body in light gray, wing doors in yellow
Specifications
TRGS 510 | |
500 x 1200 x 1950 mm | |
Storage of hazardous substances | |
2 | |
115 kg | |
Steel |
460 x 1195 x 1845 mm | |
Wing | |
300 kg | |
Powder-coated | |
Yellow | |
Safety storage cabinet |