missing translation for 'onlineSavingsMsg'
Learn More

Erlab™ Captair 834 Smart Filtering Storage Cabinets

These filtering chemical storage cabinets provide outstanding filtration quality by ensuring that users and laboratory technicians are protected against chemical hazards. Their technology adapts to the type of product being stored. Users are given the best possible view of the stored products through their shelves, transparent doors and are alerted in the event of prolonged opening.

Brand:  Erlab™ AB41//03

Additional Details : Weight : 104.00000kg

 View more versions of this product

Product Code. 17901702

  • £2994.57 / Each
Estimated Shipment: 01-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Air Flow: 220 m3/hr. (129 CFM)
  • Structure made of corrosion resistant electro-galvanized steel coated with anti-acid polymer
  • Doors made of clear, chemical resistant acrylic for easy viewing
  • Filtration module is made with injected polypropylene
  • Filtration technology: 1 column can be configured to handle liquids, powders, or both
  • Following filtration column configuration: Carbon filtration for gases and vapors and particulate filtration for powders
  • Detection sensor for Type S, for solvents; Type A, for acids; Type F, for formaldehydes
  • Door sensor alarm deploys if doors are left open
  • 2 storage compartments
  • Absorbing mats: 2
  • Key lock
  • List of approved chemicals
  • Communication Interface: When the light is pulsing: doors left open, containment is compromised, filter breakthrough
  • eGuard APP: For remote control to monitor the status of the storage cabinet, change the settings, and deliver safety alerts immediately to your devices
Specifications

Specifications

Chemicals Cabinet
35.4 in.
Integrated Retention Tray, Storage for 100 x 1 L Glass Bottles
26 in.
Adjustable
10
CE, AFNOR NF X 15 211:2009, EN 1822:1998
84.3 in.
Steel, Polymer
660 x 900 x 2140 mm
White
Swing
2
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers