missing translation for 'onlineSavingsMsg'
Learn More

BTZO 1, Tocris Bioscience™

Product Code. 18065232
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

MIF binder

Macrophage migration inhibitory factor (MIF) binder (Kd = 68.6 nM). Activates antioxidant response element (ARE)-mediated gene expression. Suppresses oxidative stress-induced cardiomyocyte apoptosis in vitro. Cardioprotective.

Chemical Identifiers

CAS 99420-15-2
Molecular Formula C13H8N2OS
Molecular Weight (g/mol) 240.28
InChI Key GBAKVEWPYUIGHN-UHFFFAOYSA-N
Synonym btzo 1, 2-2-pyridyl-4h-1,3-benzothiazin-4-one, 2-pyridin-2-yl-4h-benzo e 1,3 thiazin-4-one, 2-pyridin-2-yl-4h-1,3-benzothiazin-4-one, 2-pyridin-2-yl-1,3-benzothiazin-4-one, btzo-1, btzo hplc, 2-2-pyridinyl-4h-1,3-benzothiazin-4-one
PubChem CID 9837640
IUPAC Name 2-pyridin-2-yl-1,3-benzothiazin-4-one
SMILES C1=CC=C2C(=C1)C(=O)N=C(S2)C3=CC=CC=N3

Specifications

Quantity 10 mg
Formula Weight 240.28
Percent Purity >99%
Chemical Name or Material BTZO 1
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'