Learn More
Edulab™ Boyle'S Law Apparatus Mounted on a Sturdy Perspex Board
Brand:  Edulab™ 251-401
Code : 50
Additional Details : Weight : 1.00000kg
Product Code. 13182840
  • £121.76 / Each
Estimated Shipment: 10-09-2024
to see stock.
Add to basket
Description
Description
with gauge and syringe connected by a high pressure hose demonstrates Boyle's law white/blue Edu-Lab